Learn More
Aviva Systems Biology Corp NPAS1 Antibody - C-terminal region (ARP32053_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32053_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPAS1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NPAS1. Alternate names: MOP5, PASD5, bHLHe11ApplicationIHC, WB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52852