Learn More
Aviva Systems Biology Corp Npas2 Antibody - N-terminal region (ARP37865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Npas2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human Npas2. Alternate names: MGC129355, MOP4, bHLHe9ApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-695