Learn More
Lonza Walkersville NPBM Neural Prog Basal Medium
Catalog No. 5000496393
Encompass
Poietics Neural Progenitor Cell System contains normal human neural progenitor cells (NHNP) and defined medium optimized for their growth and in vitro differentiation This system may be used for experimental applications including drug development neurotoxicity neurogenesis electrophysiology neurotransmitter disorders and central nervous system function The available defined neural progenitor media includes a growth medium BulletKit (CC-3209) as well as a differentiaion medium BulletKit (CC-3229) Poietics Cells Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system
Catalog No. 50-004-96393 Supplier Lonza Walkersville Supplier No. CC3210
Poietics Neural Progenitor Cell System contains normal human neural progenitor cells (NHNP) and defined medium optimized for their growth and in vitro differentiation This system may be used for experimental applications including drug development neurotoxicity neurogenesis electrophysiology neurotransmitter disorders and central nervous system function The available defined neural progenitor media includes a growth medium BulletKit (CC-3209) as well as a differentiaion medium BulletKit (CC-3229) Poietics Cells Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system