Learn More
Aviva Systems Biology Corp NPBWR2 Antibody - N-terminal region (ARP65592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPBWR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human NPBWR2. Alternate names: GPR8ApplicationWB. Species reactivity: Human
Catalog No. 50-288-524