Learn More
Abcam NPC-15437, PKC inhibitor, 5MG
Catalog No. 502705583
Encompass_Preferred
MW 511.6 Da, Purity >98%. PKC inhibitor (IC₅₀ = 19 μM). Interacts with regulatory domain at residues 12-42. Neuroprotective agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5583 Supplier Abcam Supplier No. AB1446005MG
MW 511.6 Da, Purity >98%. PKC inhibitor (IC₅₀ = 19 μM). Interacts with regulatory domain at residues 12-42. Neuroprotective agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement