Learn More
Perkin Elmer US LLC NPD with PPC Add-On Kit, 240 V
Catalog No. 502113721
Encompass_Preferred
Kit includes: detector, heater, sensor, heater block, and programmable pneumatic control for detector combustion gases and two beads. Requires, but does not include, amplifier (N6120094). The AutoSystem XL or Clarus GC must be PPC ready. If not, the PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.
Catalog No. 50-211-3721 Supplier Perkin Elmer US LLC Supplier No. N6120127
Add to Cart
Add to Cart
Request a Quote
Kit includes: detector, heater, sensor, heater block, and programmable pneumatic control for detector combustion gases and two beads. Requires, but does not include, amplifier (N6120094). The AutoSystem XL or Clarus GC must be PPC ready. If not, the PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.