Learn More
Aviva Systems Biology Corp NPDC1 Antibody - N-terminal region (ARP47153_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47153_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NPDC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NPDC1. Alternate names: CAB, CAB-, CAB-1, CAB1, DKFZP586J0523, NPDC-1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat, Yeast
Catalog No. 50-014-59475