Learn More
Cayman Chemical N-pentadecanoyl-L-Homoserine lactone, Purity: ≥98%, CAS: 182359-66-6, 50 mg
SDP
Catalog No. 502953230
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; activates the LasR quorum sensing receptor of bacteria in a reporter assay; induces GFP production in a P. putida F117 reporter strain expressing the pKR-C12 quorum-sensing operon at 0.094 nM
Catalog No. 50-295-3230 Supplier Cayman Chemical Supplier No. 1309450 MG
A bacterial quorum-sensing signaling molecule; activates the LasR quorum sensing receptor of bacteria in a reporter assay; induces GFP production in a P. putida F117 reporter strain expressing the pKR-C12 quorum-sensing operon at 0.094 nM