Learn More
Cayman Chemical N-Peptide (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502916117
Encompass_Preferred
An E. coli SurA-binding protein; binds to the E. coli periplasmic molecular chaperone SurA (Kds = 2.3 and 1.69 µM for the full-length protein and core domain, respectively)
Catalog No. 50-291-6117 Supplier Cayman Chemical Supplier No. 374715 MG
An E. coli SurA-binding protein; binds to the E. coli periplasmic molecular chaperone SurA (Kds = 2.3 and 1.69 µM for the full-length protein and core domain, respectively)