Learn More
Aviva Systems Biology Corp NPFFR2 Antibody - middle region (ARP59569_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPFFR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NPFFR2. Alternate names: GPR74, HLWAR77, NPFF2, NPGPRApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-286-743