Learn More
Research Products International Corp NPG [4-Nitrophenyl-B-D-glucuronide], 1 Gram
SDP
Catalog No. 50213529
Encompass_Preferred
Spectrophotometric substrate for detection of B-D-Glucuronidase.
Catalog No. 50-213-529 Supplier Research Products International Corp Supplier No. N120301.0
Spectrophotometric substrate for detection of B-D-Glucuronidase.