Learn More
Cayman Chemical N-phenylacetyl-L-Homoserine lactone, Purity: ≥98%, CAS: 55154-48-8, 10 mg
SDP
Catalog No. 502892633
Encompass_Preferred
A small diffusible signaling molecule involved in quorum sensing in the model symbiont V. fischeri and the pathogens A. tumefaciens and P. aeruginosa
Catalog No. 50-289-2633 Supplier Cayman Chemical Supplier No. 900173710 MG
A small diffusible signaling molecule involved in quorum sensing in the model symbiont V. fischeri and the pathogens A. tumefaciens and P. aeruginosa