Learn More
Cayman Chemical N-Phenylacetyl-L-Prolylglycine ethyl ester, Purity: ≥98%, CAS: 157115-85-0, 10 mg
SDP
Catalog No. 502970976
Encompass_Preferred
A synthetic dipeptide that has been shown to produce positive nootropic and cognitive effects in animal models (0.01-0.8 mg/kg) by a mechanism similar to other related racetam compounds; intended for research and forensic purposes only
Catalog No. 50-297-0976 Supplier Cayman Chemical Supplier No. 1449610 MG
A synthetic dipeptide that has been shown to produce positive nootropic and cognitive effects in animal models (0.01-0.8 mg/kg) by a mechanism similar to other related racetam compounds; intended for research and forensic purposes only