Learn More
KINOVATE LIFE SCIENCES INC Kinovate Life Sciences Solid Support, NittoPhase HL, NPHL 2'OMeA(bz), High-Loading
Catalog No. NC3973595
Encompass
NittoPhase HL is a high-loading solid support for therapeutic oligonucleotide synthesis, building on the foundation of the product line which has been in use since 2004. It enables higher synthesis scales per column volume while maintaining superior yields and full-length purity. This product allows for efficiency and cost reduction in oligonucleotide synthesis without compromising quality.
  • Loading capacity: up to 400 μmol/g for DNA & RNA synthesis.
  • Synthesis scale: proven performance in commercial production at scales up to 400 mmol.
  • Manufactured under ISO 9001:2015 guidelines.
  • Flexible for synthesizing RNA, longmers, and other modified molecules at higher loadings.
  • Consistent high yields and improved purity profiles across various sequences.
Catalog No. NC3973595 Supplier KINOVATE LIFE SCIENCES INC Supplier No. 241200350
NittoPhase HL is a high-loading solid support for therapeutic oligonucleotide synthesis, building on the foundation of the product line which has been in use since 2004. It enables higher synthesis scales per column volume while maintaining superior yields and full-length purity. This product allows for efficiency and cost reduction in oligonucleotide synthesis without compromising quality.
  • Loading capacity: up to 400 μmol/g for DNA & RNA synthesis.
  • Synthesis scale: proven performance in commercial production at scales up to 400 mmol.
  • Manufactured under ISO 9001:2015 guidelines.
  • Flexible for synthesizing RNA, longmers, and other modified molecules at higher loadings.
  • Consistent high yields and improved purity profiles across various sequences.