Learn More
Aviva Systems Biology Corp NPHP1 Antibody - middle region (ARP56084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPHP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NPHP1. Alternate names: JBTS4, NPH1, SLSN1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61712