Learn More
Aviva Systems Biology Corp NPIP Antibody - middle region (ARP63842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPIP. It was validated on Western Blot. . Alternate names: FLJ42525, FLJ44848, morpheus, NPIPApplicationWB. Species reactivity: Human
Catalog No. 50-288-097