Learn More
Sino Biological Recombinant Human ALK Protein (L1196M, G1202R, GST Tag), Active
Catalog No. 502575945
Encompass_Preferred
Recombinant human ALK (L1196M G1202R) (1060-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5945 Supplier Sino Biological Supplier No. A1912NG50
Recombinant human ALK (L1196M G1202R) (1060-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.