Learn More
Aviva Systems Biology Corp NPM1 Antibody - middle region (ARP34095_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34095_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NPM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NPM1. Alternate names: B23, NPMApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55246