Learn More
Aviva Systems Biology Corp NPM1 Antibody - C-terminal region (ARP34114_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34114_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NPM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NPM1. Alternate names: B23, NPMApplicationIHC, WB. Species reactivity: Human, Mouse
Catalog No. 50-014-55253