Learn More
Medchemexpress LLC NPR2 Human P. past 2ug
SDP
Catalog No. 5000182858
Encompass_Preferred
NPR2 Protein Human (P pastoris His) serves as the receptor for C-type natriuretic peptide NPPC/CNP activating guanylate cyclase upon ligand binding NPR2 potentially regulates skeletal growth indicating involvement in bone development Its interaction with C-type natriuretic peptide highlights its significance in mediating cellular responses especially in signaling pathways influencing skeletal growth dynamics NPR2 Protein Human (P pastoris His) is the recombinant human-derived Atrial natriuretic peptide receptor 2/NPR2 expressed by P pastoris with N-His labeled tag
Catalog No. 50-001-82858 Supplier Medchemexpress LLC Supplier No. HYP723032UG
NPR2 Protein Human (P pastoris His) serves as the receptor for C-type natriuretic peptide NPPC/CNP activating guanylate cyclase upon ligand binding NPR2 potentially regulates skeletal growth indicating involvement in bone development Its interaction with C-type natriuretic peptide highlights its significance in mediating cellular responses especially in signaling pathways influencing skeletal growth dynamics NPR2 Protein Human (P pastoris His) is the recombinant human-derived Atrial natriuretic peptide receptor 2/NPR2 expressed by P pastoris with N-His labeled tag