Learn More
Apexbio Technology LLC NPS-1034 1221713-92-3 10mg
SDP
Catalog No. 5000088330
Encompass_Preferred
NPS-1034 (CAS 1221713-92-3) is a potent inhibitor of the receptor tyrosine kinase MET with an IC50 of 4 nmol/L MET activated by hepatocyte growth factor is implicated in morphogenesis and tumor progression across various cancers NPS-1034 suppresses cell viability in MET-overexpressing and phosphorylated MET-positive cell lines such as MKN45 and SNU638 with IC50 values of 112 7 nmol/L and 190 3 nmol/L respectively In vivo oral administration of NPS-1034 reduces tumor growth and angiogenesis in MKN45 xenograft models These characteristics support the utility of NPS-1034 in studying MET-driven oncogenic signaling and its therapeutic targeting
Catalog No. 50-000-88330 Supplier Apexbio Technology LLC Supplier No. B600610
NPS-1034 (CAS 1221713-92-3) is a potent inhibitor of the receptor tyrosine kinase MET with an IC50 of 4 nmol/L MET activated by hepatocyte growth factor is implicated in morphogenesis and tumor progression across various cancers NPS-1034 suppresses cell viability in MET-overexpressing and phosphorylated MET-positive cell lines such as MKN45 and SNU638 with IC50 values of 112 7 nmol/L and 190 3 nmol/L respectively In vivo oral administration of NPS-1034 reduces tumor growth and angiogenesis in MKN45 xenograft models These characteristics support the utility of NPS-1034 in studying MET-driven oncogenic signaling and its therapeutic targeting