Learn More
Medchemexpress LLC NPS-1034 | 1221713-92-3 | 98.0% | 551.54 | C31H23F2N5O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10050910MG
Encompass_Preferred
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET. It exhibits IC50 values of 10.3 nM (AXL) and 48 nM (MET) and is supplied for research use as a solid powder or as a 10 mM solution in DMSO.
  • Dual inhibition of AXL and MET with low-nanomolar potency.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Available as solid powder or DMSO solution for assay flexibility.
  • Supplied in multiple package sizes to fit experimental needs.
  • Useful for studying MET/AXL signaling and drug resistance mechanisms.
Catalog No. 50-002-14811