Learn More
Medchemexpress LLC NPS-1034 | 1221713-92-3 | 98.0% | 551.54 g/mol | C31H23F2N5O3 | 1 ML
SDP
Catalog No. 5000214149
Encompass_Preferred
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET, exhibiting low-nanomolar biochemical potency (AXL IC50 = 10.3 nM; MET IC50 = 48 nM). It is supplied for research use in formats including solid powder and ready-to-use 10 mM solution in DMSO, and is characterized by CAS 1221713-92-3 and molecular weight 551.54 g/mol.
  • Dual inhibition of AXL and MET with low-nanomolar potency.
  • Suitable for in vitro kinase and cell-based studies.
  • Available as solid powder or 10 mM DMSO solution for convenience.
  • Characterized by CAS 1221713-92-3 and molecular weight 551.54 g/mol.
  • High purity (98.0%) for consistent experimental results.
Catalog No. 50-002-14149 Supplier Medchemexpress LLC Supplier No. HY10050910MM 1ML
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET, exhibiting low-nanomolar biochemical potency (AXL IC50 = 10.3 nM; MET IC50 = 48 nM). It is supplied for research use in formats including solid powder and ready-to-use 10 mM solution in DMSO, and is characterized by CAS 1221713-92-3 and molecular weight 551.54 g/mol.
  • Dual inhibition of AXL and MET with low-nanomolar potency.
  • Suitable for in vitro kinase and cell-based studies.
  • Available as solid powder or 10 mM DMSO solution for convenience.
  • Characterized by CAS 1221713-92-3 and molecular weight 551.54 g/mol.
  • High purity (98.0%) for consistent experimental results.