Learn More
Medchemexpress LLC Nps-1034 | 1221713-92-3 | 98.0% | 551.54 g/mol | C31H23F2N5O3 | 50 MG
SDP
Catalog No. 5000214147
Encompass_Preferred
NPS-1034 is a small-molecule, research-use inhibitor targeting the receptor tyrosine kinases AXL and MET. It inhibits AXL with an IC50 of 10.3 nM and MET with an IC50 of 48 nM, and is supplied as a powder for laboratory studies to probe MET/AXL signaling and evaluate anti-tumor activity.
  • Potent dual kinase inhibition: AXL IC50 10.3 nM, MET IC50 48 nM.
  • High purity suitable for research (98.0%).
  • Supplied as powder for flexible dosing and formulation.
  • DMSO solubility approximately 34 mg/mL; ultrasonic assistance recommended.
  • Storage stability: powder stable at -20°C for long-term storage.
Catalog No. 50-002-14147 Supplier Medchemexpress LLC Supplier No. HY10050950MG
NPS-1034 is a small-molecule, research-use inhibitor targeting the receptor tyrosine kinases AXL and MET. It inhibits AXL with an IC50 of 10.3 nM and MET with an IC50 of 48 nM, and is supplied as a powder for laboratory studies to probe MET/AXL signaling and evaluate anti-tumor activity.
  • Potent dual kinase inhibition: AXL IC50 10.3 nM, MET IC50 48 nM.
  • High purity suitable for research (98.0%).
  • Supplied as powder for flexible dosing and formulation.
  • DMSO solubility approximately 34 mg/mL; ultrasonic assistance recommended.
  • Storage stability: powder stable at -20°C for long-term storage.