Learn More
Selleck Chemical LLC NPS-1034 5mg 1221713-92-3
SDP
Catalog No. 501365103
Encompass_Preferred
NPS-1034 is a dual Met (c-Met)/Axl inhibitor with IC50 of 48 nM and 10.3 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5103 Supplier Selleck Chemical LLC Supplier No. S76695MG
NPS-1034 is a dual Met (c-Met)/Axl inhibitor with IC50 of 48 nM and 10.3 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.