Learn More
Medchemexpress LLC HY-100509 5mg Medchemexpress, NPS-1034 CAS:1221713-92-3 Purity:>98%
SDP
Catalog No. 501931860
Encompass_Preferred
Medchemexpress, HY-100509 5mg NPS-1034 CAS:1221713-92-3 NPS-1034 is a dual inhibitor of AXL and MET with IC 50 s of 10.3 and 48 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1860 Supplier Medchemexpress LLC Supplier No. HY1005095MG
Medchemexpress, HY-100509 5mg NPS-1034 CAS:1221713-92-3 NPS-1034 is a dual inhibitor of AXL and MET with IC 50 s of 10.3 and 48 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.