Learn More
Selleck Chemical LLC NPS-2143-10mg
SDP
Catalog No. 502390003
Encompass_Preferred
NPS-2143 (SB262470) is a novel potent and selective antagonist of Ca(2+) receptor with IC50 of 43 nM.
Catalog No. 50-239-0003 Supplier Selleck Chemical LLC Supplier No. S263310MG
NPS-2143 (SB262470) is a novel potent and selective antagonist of Ca(2+) receptor with IC50 of 43 nM.