Learn More
Medchemexpress LLC HY-10007 100mg , NPS-2143 SB 262470A CAS:284035-33-2 Purity:98%
SDP
Catalog No. 501493005
Encompass_Preferred
Medchemexpress, HY-10007 100mg NPS-2143 SB 262470A CAS:284035-33-2 Formula:C24H25ClN2O2 IC50: 43 nM (Ca 2+ receptor) Purity:98% NPS-2143 is a novel potent and selective antagonist of  Ca 2+ receptor  with  IC 50  of 43 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3005 Supplier Medchemexpress LLC Supplier No. HY10007100MG
Medchemexpress, HY-10007 100mg NPS-2143 SB 262470A CAS:284035-33-2 Formula:C24H25ClN2O2 IC50: 43 nM (Ca 2+ receptor) Purity:98% NPS-2143 is a novel potent and selective antagonist of  Ca 2+ receptor  with  IC 50  of 43 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.