Learn More
Medchemexpress LLC HY-10171 5mg Medchemexpress, NPS-2143 (hydrochloride) CAS:324523-20-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101715MG
Encompass_Preferred
Medchemexpress, HY-10171 5mg NPS-2143 (hydrochloride) CAS:324523-20-8 NPS-2143 hydrochloride (SB-262470A hydrochloride), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 hydrochloride (SB-262470A hydrochloride) blocks increases in cytoplasmic Ca 2+ concentrations ( IC 50 =43 nM) elicited by activating the Ca 2+ receptor in HEK 293 cells expressing the human Ca 2+ receptor [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2196