Learn More
Medchemexpress LLC NPS-2143 hydrochloride | 324523-20-8 | MFCD04113010 | 100.0% | 445.38 | C24H26Cl2N2O2 | 1 ML
SDP
Catalog No. 5000245647
Encompass_Preferred
NPS-2143 hydrochloride is a selective and potent calcium-sensing receptor (CaSR) antagonist used as a calcilytic research tool to modulate intracellular calcium signaling and parathyroid hormone secretion. It is supplied as a white to off-white solid and is available pre-formulated as a 10 mM solution in DMSO for laboratory use.
  • Selective calcium-sensing receptor antagonist (IC50 = 43 nM).
  • Useful for studies of CaSR signaling and parathyroid hormone release.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • White to off-white solid form for easy handling and storage.
Catalog No. 50-002-45647 Supplier Medchemexpress LLC Supplier No. HY1017110MM 1ML
NPS-2143 hydrochloride is a selective and potent calcium-sensing receptor (CaSR) antagonist used as a calcilytic research tool to modulate intracellular calcium signaling and parathyroid hormone secretion. It is supplied as a white to off-white solid and is available pre-formulated as a 10 mM solution in DMSO for laboratory use.
  • Selective calcium-sensing receptor antagonist (IC50 = 43 nM).
  • Useful for studies of CaSR signaling and parathyroid hormone release.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • White to off-white solid form for easy handling and storage.