Learn More
Medchemexpress LLC NPS-2143 hydrochloride | 324523-20-8 | MFCD16038896 | 100.0% | 445.4 g/mol | C24H26Cl2N2O2 | 1 MG
SDP
Catalog No. 5000246996
Encompass_Preferred
NPS-2143 hydrochloride is an orally active calcilytic agent that acts as a selective and potent antagonist of the calcium-sensing receptor (CaSR). Supplied as a high-purity solid for research use, it is used in biochemical and cellular studies to probe CaSR-mediated signaling and parathyroid hormone regulation.
  • Selective calcium-sensing receptor antagonist for targeted research applications.
  • High purity suitable for biochemical and cellular assays.
  • Solid form for convenient storage and handling.
  • Small research-scale quantity for assay development and proof-of-concept studies.
Catalog No. 50-002-46996 Supplier Medchemexpress LLC Supplier No. HY101711MG
NPS-2143 hydrochloride is an orally active calcilytic agent that acts as a selective and potent antagonist of the calcium-sensing receptor (CaSR). Supplied as a high-purity solid for research use, it is used in biochemical and cellular studies to probe CaSR-mediated signaling and parathyroid hormone regulation.
  • Selective calcium-sensing receptor antagonist for targeted research applications.
  • High purity suitable for biochemical and cellular assays.
  • Solid form for convenient storage and handling.
  • Small research-scale quantity for assay development and proof-of-concept studies.