Learn More
Medchemexpress LLC NPS-2143 hydrochloride | 324523-20-8 | MFCD16038896; MFCD04113010 | 100.0% | 445.38 g/mol | C24H26Cl2N2O2 | 50 MG
SDP
Catalog No. 5000248438
Encompass_Preferred
NPS-2143 hydrochloride is an orally active calcilytic small molecule that selectively antagonizes the calcium-sensing receptor (CaSR), inhibiting CaSR-mediated increases in intracellular Ca2+. It is supplied as a solid at high purity for use in biochemical and pharmacological research into CaSR signaling and related physiological pathways.
  • Selective CaSR antagonist for CaSR signaling studies.
  • Orally active calcilytic small molecule suitable for in vitro and in vivo research.
  • High purity solid appropriate for research applications.
  • Molecular weight 445.38 g/mol and chemical formula C24H26Cl2N2O2.
  • Available in small research quantities, including 50 MG.
  • Applicable to biochemical, cellular, and pharmacological assays.
Catalog No. 50-002-48438 Supplier Medchemexpress LLC Supplier No. HY1017150MG
NPS-2143 hydrochloride is an orally active calcilytic small molecule that selectively antagonizes the calcium-sensing receptor (CaSR), inhibiting CaSR-mediated increases in intracellular Ca2+. It is supplied as a solid at high purity for use in biochemical and pharmacological research into CaSR signaling and related physiological pathways.
  • Selective CaSR antagonist for CaSR signaling studies.
  • Orally active calcilytic small molecule suitable for in vitro and in vivo research.
  • High purity solid appropriate for research applications.
  • Molecular weight 445.38 g/mol and chemical formula C24H26Cl2N2O2.
  • Available in small research quantities, including 50 MG.
  • Applicable to biochemical, cellular, and pharmacological assays.