Learn More
Cayman Chemical NPS 2143hydrochlorIde 10mg
SDP
Catalog No. 502967928
Encompass_Preferred
A potent and selective antagonist of CaSR (IC50 = 43 nM) that stimulates PTH secretion from parathyroid cells in vitro (EC50 = 39 nM); causes a sustained increase in plasma PTH in rats when given intravenously
Catalog No. 50-296-7928 Supplier Cayman Chemical Supplier No. 1790310 MG
A potent and selective antagonist of CaSR (IC50 = 43 nM) that stimulates PTH secretion from parathyroid cells in vitro (EC50 = 39 nM); causes a sustained increase in plasma PTH in rats when given intravenously