Learn More
Medchemexpress LLC NPS 2390 | 226878-01-9 | 99.9% | 307.39 | 10 MG
SDP
Catalog No. 5000343674
Encompass_Preferred
NPS 2390 is a noncompetitive antagonist of mGluR1 and mGluR5. It is also a potent CaSR (calcium-sensing receptor) inhibitor.
  • Noncompetitive antagonist of mGluR1 and mGluR5.
  • Potent CaSR (calcium-sensing receptor) inhibitor.
  • For research use only.
Catalog No. 50-003-43674 Supplier Medchemexpress LLC Supplier No. HY11095100MG
NPS 2390 is a noncompetitive antagonist of mGluR1 and mGluR5. It is also a potent CaSR (calcium-sensing receptor) inhibitor.
  • Noncompetitive antagonist of mGluR1 and mGluR5.
  • Potent CaSR (calcium-sensing receptor) inhibitor.
  • For research use only.