Learn More
FyterTech Nonwovens MERCSORB™ Mercury Amalgamation Powder
Catalog No. 19066526 Shop All NPS Corporation Products
Change view
Click to view available options
Packaging Quantity:
500 g
Catalog No. Packaging Quantity
19-066-526 500 g
1 options
Catalog No. 19-066-526 Supplier FyterTech Nonwovens Supplier No. 520500
Add to Cart
Add to Cart
Use for dangerous mercury spills
  • Amalgamation powder converts elemental mercury into amalgam and stops dangerous mercury vapors from being emitted
  • Components: Zinc dust, Citric acid
Specifications
Material Zinc Dust, Citric Acid
Type Amalgamation Powder
For Use With (Application) Convert elemental mercury into amalgam and stops dangerous mercury vapors from being emitted, for dangerous mercury spills
Packaging Quantity 500 g
To be used by trained personnel only