Learn More
FyterTech Nonwovens Specialty Spill Control, Mini Acid/Base/Alkaline Spill Kit
Catalog No. 19066480 Shop All NPS Corporation Products
Change view
Click to view available options
Mini Acid/Base/Alkaline Spill Kit
Catalog No.
19-066-480 Mini Acid/Base/Alkaline Spill Kit
1 options
Catalog No. 19-066-480 Supplier FyterTech Nonwovens Supplier No. 271340
Add to Cart
Add to Cart
Includes
One quart bottle liquid acid neutralizer, one quart bottle liquid base/alkaline neutralizer, Five 12 x 12 in. Hazmat pads, one disposal bag and twist tie
  • This Kit takes up very little space on shelves or table tops
  • Box has removable perforation to hold contents but keep them visible before use
  • Color Change process indicates when acids or bases are safe to handle
  • 4 kits/case
  • 10.4kg (23 lb.)
Specifications
Type Mini Acid/Base/Alkaline Spill Kit
Includes Acid and base neutralizer, 5 Hazmat pads (12 x 12 in.), 1 disposal bag and twist tie