Learn More
FyterTech Nonwovens Specialty Spill Control, Odorous Liquids Spill Kit
Shop All NPS Corporation ProductsDescription
Includes
One 0.5 gal. carton odorous absorbent, One whisk on pan and sweep, one pair nitrile gloves, one disposal bag and twist tie
- Environmentally Safe and easy to use, quickly and easily absorbs fluid accidents and other odorous liquids
- Absorbs up to 10 times its own weight
- Works on all surfaces
- Absorbs 2 gal./carton or 8 gal./case
- Packed in convenient shaker carton
Specifications
Specifications
| Type | Odorous Liquids Spill Kit |
| Includes | 0.5 gal. carton odorous absorbent, 1 whisk on pan and sweep, 1 pair nitrile gloves, 1 disposal bag and twist tie |