Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Spilfyter™ HazMat Poly Drum Spill Kit

Catalog No. 19066507 Shop All NPS Corporation Products
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-066-507 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-066-507 Supplier FyterTech Nonwovens Supplier No. 250055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

3 in. x 4 ft. Socks (20), 16 x 18 in. Pads (200), Disposal Bag w/ Twist Tie (1), Nitrile Gloves (1 pr.), Splash Goggles (1 pr.), Emergency Response Guidebook (1), 55 Gallon Poly Drum (1)

Spill response materials packaged in a Poly Drum

  • Kits contained in durable Poly Drum with Lever-Lock metal ring tops
  • Contains Hazmat spill response socks and pads for aggressive or unkown solution spills
  • Designed for either inside or outside storage
  • Absorbs up to 72 gal. (272 liters) per kit

Compliance

Poly Drums are UN-Certified

Specifications

Capacity (English) 55 gal.
Capacity (Metric) 208 L
Material Polypropylene Particulate, Fabric
Type Sorbent Spill Kit
Includes 20 socks, 200 pads, 1 twist tie, 1 pair nitrile gloves, 1 pair splash goggles, 1 emergency response guidebook, 1 poly drum 55 gal.
Weight (English) 64 lb.
Weight (Metric) 29 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.