Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting Kit, Polypropylene, Elbow/Tee/Y, Hose Barb; 1 Set
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4151415/DEL
Encompass
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Catalog No. NC0981518