Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 1/8"" ID x 1/8"" NPT(F); 25/PK
Catalog No. NC1588789
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. NC1588789 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3150040
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).