Learn More
Aviva Systems Biology Corp NPTN Antibody - middle region (ARP46899_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46899_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NPTN. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NPTN. Alternate names: DKFZp686L2477, GP55, GP65, MGC102805, SDFR1, SDR1, np55, np65ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59433