Learn More
Cayman Chemical NpTosylGlyProLyspNAac 10mg
SDP
Catalog No. 502990874
Encompass_Preferred
A colorimetric plasmin substrate; hydrolysis releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasmin activity
Catalog No. 50-299-0874 Supplier Cayman Chemical Supplier No. 3154410 MG
A colorimetric plasmin substrate; hydrolysis releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasmin activity