Learn More
Aviva Systems Biology Corp NPTX2 Antibody - N-terminal region (ARP56365_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56365_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPTX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NPTX2. Alternate names: NARP, NP-II, NP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-144