Learn More
TARGETMOL CHEMICALS INC NPX800 5MG
SDP
Catalog No. 502578957
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NPX800 is an inhibitor of heat shock factor 1 (HSF1) and can be used in studies about the treatment of cancer. purity: 99%
Catalog No. 50-257-8957 Supplier TARGETMOL CHEMICALS INC Supplier No. T98045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NPX800 is an inhibitor of heat shock factor 1 (HSF1) and can be used in studies about the treatment of cancer. purity: 99%