Learn More
Enzo Life Sciences NPY (human) (1mg). CAS: 90880-35-6
SDP
Supplier:  Enzo Life Sciences ALX163003P001
Encompass_Preferred
Alternative name: Neuropeptide Y, Neuropeptide tyrosine. Formula: C189H285N55O57S. MW: 4271.7. Purity: ≥97%. Appearance: White lyophilized powder. MI: 14: 6485. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7846