Learn More
Carbosynth LLC. NPY (Porcine, 13-36) (0.1mg vial), 1ea, MW: 2982.4, Molecular formula: C135H209N41O36, CAS# 113662-54-7
SDP
Supplier:  Carbosynth LLC. PNP4315S0.1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2Neuropeptide Y, Application: NPY Y2-Receptor Selective Agonist, Storage Temperature: -20C, References: M.W. Walker and R.J. Miller, Mol. Pharmacol., 34, 779 (1988). (Pharmacol.), J. Fuhlendorff, U. Gether, L. Aakerlund, N. Langeland-Johansen, H.Thgersen, S.G. Melberg, U.B. Olsen, O. Thastrup, and T.W. Schwartz, Proc. Natl. Acad. Sci. USA, 87, 182 (1900). (Pharmacol.), S.P. Sheikh, Am. J. Physiol., 261, G701 (1991). (Pharmacol.)
Catalog No. 50-168-6704