Learn More
Aviva Systems Biology Corp NPY1R Antibody - middle region (ARP41827_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPY1R. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NPY1R. Alternate names: NPYR, NPY1-RApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58009