Learn More
Enzo Life Sciences Neuropeptide Y receptor Y1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905798100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat neuropeptide Y receptor Y1. UniProt ID: P21555. Species reactivity: Human, Mouse, Rat. Creossreactivity: Predicted species reactivity based on epitope sequence homology: horse, chimpanzee, rhesus macaque, guinea pig. Applications: ELISA, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-200-7063