Learn More
MyBioSource NPYR2 (Human) ELISA Kit, Neuropeptide Y Receptor Y2
SDP
Supplier:  MyBioSource MBS76291996STRIPWELLS
Encompass_Preferred
The MyBioSource Human Neuropeptide Y Receptor Y2 ELISA Kit quantifies NPYR2 in various human biological fluids using a 96-well plate coated with a specific antibody. Samples and standards are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and TMB substrate. The color change, measured at 450 nm, indicates NPYR2 concentration.
Catalog No. NC2201352