Learn More
Eurofins Discoverx PathHunter NPY2R Activated GPCR Internalization U2OS Cell Line, 2 vials. (93-0721C3)
Catalog No. 502001187
Encompass
The PathHunter NPY2R Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of NPY2R via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged NPY2R. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting NPY2R-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1187 Supplier Eurofins Discoverx Supplier No. 930721C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter NPY2R Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of NPY2R via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged NPY2R. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting NPY2R-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.